Key Documents
Z708224
Well Pads
48 well
About This Item
UNSPSC Code:
41122102
sterility
non-sterile
manufacturer/tradename
Diversified Biotech WPNP-2000
size
48 wells
color
none
General description
A note pad that corresponds to the layout of a 96-, 48- or 24-well plate. A fast and easy way to keep accurate records of samples in a microplate. The adhesive backing allows the researcher to affix individual sheets into a notebook. The pad contains (50) 5" × 7" sheets.
Choose from one of the most recent versions:
Certificates of Analysis (COA)