Key Documents
Z708097
Sigma-Aldrich
Lab tables
Small
About This Item
UNSPSC Code:
41100000
W × H × D
8.75 in. × 11.75 in. × 7.75 in.
General description
Now you can raise components off your bench and free up your workspace. These handy items are made from strong 1/2in. PVC. Units of same width are stackable. Great for stacking electrical components. Holes for running cords
Choose from one of the most recent versions:
Certificates of Analysis (COA)