Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z706108

Spectrafuge Mini replacement rotor 6 x 1.5 mL

pkg of 1 ea

Synonym(s):

equipment replacement accessory

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$52.20

$52.20


Estimated to ship onMay 31, 2025



Select a Size

Change View
1 EA
$52.20

About This Item

UNSPSC Code:
41103900
NACRES:
NB.22

$52.20


Estimated to ship onMay 31, 2025


packaging

pack of 1 ea
pkg of 1 ea

manufacturer/tradename

Labnet C1300-RT

technique(s)

centrifugation: suitable

compatibility

for use with centrifuge models Spectrafuge mini-centrifuge

Related Categories

Legal Information

Spectrafuge is a trademark of Labnet Intl., Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service