Z692638
microDOC Compact gel documentation system
with TotalLab™ 1D software, AC/DC input 110 V AC
All Photos(1)
About This Item
UNSPSC Code:
41105300
NACRES:
NB.22
manufacturer/tradename
Cleaver CSL-MICRODOC1D$
General description
The microDOC Compact Gel Documentation System gives you all of the advantages of the Basic system with the advantage of a large 8 inch TFT colour monitor allowing clear and simple image visualization. Can be used with most 12 x 12 cm tranilluminators.
Available with optional TotalLab™ 1D software. Main software functions include: lane creation, background subtraction, band detection, profile deconvolution, and molecular size, pI and quantity calibration.
System comprises of:
Canon G12 digital
2MB storage card
Ethidium bromide & close up +3 filters
Lift-off hood with easy access door and built-in inner lights
8 in. TFT color monitor
Available with optional TotalLab™ 1D software. Main software functions include: lane creation, background subtraction, band detection, profile deconvolution, and molecular size, pI and quantity calibration.
System comprises of:
Canon G12 digital
2MB storage card
Ethidium bromide & close up +3 filters
Lift-off hood with easy access door and built-in inner lights
8 in. TFT color monitor
Legal Information
TotalLab is a trademark of Nonlinear Dynamics Ltd.
Choose from one of the most recent versions:
Certificates of Analysis (COA)