Key Documents
Z692581
microDOC Basic gel documentation system
with TotalLab 1D software, AC/DC input 230 V AC
About This Item
UNSPSC Code:
41105300
NACRES:
NB.22
feature
contains UK and Schuko plugs
manufacturer/tradename
Cleaver CSL-MDOCBASIC1D
General description
This is the perfect low cost option to upgrade to digital documentation of your gels. Can be used with most 21 x 21 cm tranilluminators.
Main software functions include: lane creation, background subtraction, band detection, profile deconvolution, and molecular size, pI and quantity calibration.
System comprises of:
  • Canon G15 digital camera (12.1 MP)
  • 2GB storage card
  • Ethidium bromide & close up +3 filters
  • Lift-off hood
  • Available with optional TotalLab 1D software
Legal Information
TotalLab is a trademark of Nonlinear Dynamics Ltd.
Choose from one of the most recent versions:
Certificates of Analysis (COA)