Key Documents
Z692085
Syringe pump connector kit, for 3DKUBE 3D cell culture chambers
pkg of 6 ea
Synonym(s):
3D, 3D Cell Culture
About This Item
UNSPSC Code:
41100000
packaging
pkg of 6 ea
manufacturer/tradename
Kiyatec FC030
General description
Syringe Pump Connector Kit provides connection between 3DKUBE and syringe or reservoir. Each connector kit contains two (2) tubing assemblies consisting of assembled tubes and Luer fittings.
Each 3DKUBE utilizes either one (1) or two (2) kits depending on configuration. Kits are provided in a convenient pouch and are gamma irradiated. Each case contains six (6) connector kits.
Legal Information
3DKUBE is a trademark of Kiyatec, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Karolina Janeczek Portalska et al.
Journal of tissue engineering and regenerative medicine, 10(5), 363-373 (2013-04-18)
Modular tissue engineering is a strategy to create scalable, self-assembling, three-dimensional (3D) tissue constructs. This strategy was used to deliver endothelial-like cells derived from bone marrow mesenchymal stromal cells (EL-MSCs) to locally induce vascularization. First, tissue engineered modules were formed
Sayed-Hadi Mirmalek-Sani et al.
Biomaterials, 34(22), 5488-5495 (2013-04-16)
Emergent technologies of regenerative medicine have the potential to overcome the limitations of organ transplantation by supplying tissues and organs bioengineered in the laboratory. Pancreas bioengineering requires a scaffold that approximates the biochemical, spatial and vascular relationships of the native