Z689513
Jenco™ micrometer for Jenco™ compound microscopes
10 mm scale with 100 divisions
All Photos(1)
About This Item
UNSPSC Code:
41191506
manufacturer/tradename
Jenco
General description
You can measure dimensions of the sample within the field of view by inserting one of the Jenco micrometers or reticules into the eyepiece. The reticules are available in multiple configurations to satisfy most needs.
Use the stage micrometer to calibrate the reticule. Its 1 mm scale is divided into 100 divisions.
Use the stage micrometer to calibrate the reticule. Its 1 mm scale is divided into 100 divisions.
Legal Information
Jenco is a trademark of Jenco International, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)