Key Documents
Z689505
Jenco micrometer for Jenco stereo microscopes
cross hair with 10 mm scale, 100 divisions
About This Item
UNSPSC Code:
41191506
manufacturer/tradename
Jenco
General description
You can measure dimensions of the sample within the field of view by inserting one of the Jenco micrometers or reticules into the eyepiece. The reticules are available in multiple configurations to satisfy most needs.
Use the stage micrometer to calibrate the reticule. Its 1 mm scale is divided into 100 divisions.
Legal Information
Jenco is a trademark of Jenco International, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)