Key Documents
Z689408
Jenco dual gooseneck light guide for Jenco illuminator
self-supporting, flexible, L 18 in.
About This Item
UNSPSC Code:
41100000
NACRES:
MA.44
manufacturer/tradename
Jenco
L
18 in.
General description
The independent illuminators (Z689386) have a 150 or 30 Watt variable intensity halogen light source. Three different light guides are available to attach to this source: a flexible single gooseneck (Z689394), a bifurcated or dual gooseneck (Z689408) and a ring light (Z689416). The 30 Watt source will function only with a single gooseneck.
  • Goosenecks direct light to any angle or position and once positioned, remain in place
  • Focusing Lens is available for attaching to the end of the gooseneck to increase intensity and improve spotlighting
  • The Ring Light attaches to the microscope shroud and moves with the optical body, eliminating the need for adjustment
  • Polarizer and analyzer are available for the ring light
Choose from one of the most recent versions:
Certificates of Analysis (COA)