Z689076
Jenco™ stereo zoom microscope, GL6 series
binocular, 10X - 65X magnification, dual halogen, AC/DC input 220 V AC, UK plug
About This Item
manufacturer/tradename
Jenco
General description
The comfort design allows for long duration tasks making them less fatiguing with accessible controls. Bonders and probers provide a compact design, and a long working distance, and the microscopes are fully integrated with camera and software systems.
All models include: a pair of 10X eye pieces, black/white reversible stage plate, dust cover, power cord and instruction manual.
- Wide interpupillary adjustment (45 - 75mm) and diopters built-in to the eyepieces
- Side mounted zoom controls and low positioned focus control with tension adjustment
- The clear, non-distorted 3D image resolved by the GL Series optics is a result of advanced technologies in optical design.
- A 100 mm working distance
- Large zoom range gives accurate reproduction of the sample up to 260X
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)