Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z688649

Nunc® Nunclon Vita Multidish, 6 well

polystyrene, flat bottom, 9.6 cm2/well, sterile, 4/cs

Synonym(s):

cell culture multidish, cell culture multiwell dish, tissue culture multidish, tissue culture multiwell dish

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121812

material

polystyrene
rectangular

description

Nunclon Vita surface treatment

sterility

sterile

feature

lid: no

packaging

case of 4 ea

manufacturer/tradename

Nunc 145380

H

58 mm

dish L × W

128 mm × 86 mm

dish capacity

18 mL

size

6 wells

surface area

9.6 cm2 , culture area (cm2) per well

well working volume

3 mL

suitability

suitable for (supports attachment, colony formation and growth of human ESC and human IPS cells)

General description

Nunclon Vita Surface supports attachment, colony formation and growth of human ESC and human IPS cells in the absence of feeder cells and matrix coatings. In media supplemented with ROCK inhibitor, human ESC can be cultured on the Nunclon Vita Surface for at least 10 passages without loss of pluripotency. Human ESC has successfully been expanded for more than 10 passages without changes to the karyotype.

Nunclon Vita is a unique energy-treated polystyrene surface that enables growth of most cell lines without using matrix coatings or feeder cells. Eliminating the need for coatings and feeder cells removes variability, requires less work, and allows for the treated surface to be adaptable to scalable cell expansion. The surface works in synergy with a ROCK inhibitor for expansion of human pluripotent stem cells and HEK cells.

  • Animal component-free surface for growth of human stem cells
  • No coating or feeder cells necessary
  • Supports attachment and colony formation of human ESC and human IPS
  • Supports growth of pluripotent human ESC for at least 10 passages
  • Non-enzymatic release and dissociation with removal of ROCK inhibitor

Legal Information

Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service