Key Documents
Z688347
LightSafe 100-place freezer storage box
black polypropylene
About This Item
UNSPSC Code:
41123300
NACRES:
NB.22
material
black polypropylene
manufacturer/tradename
Cole Parmer 675531
technique(s)
cryopreservation: suitable
L × W × H
14 cm × 14 cm × 5.4 cm
General description
The LightSafe freezer storage box is ideal for light-sensitive specimens, this all black storage box is suitable for use with 0.5, 1.5, and 2.0 mL micro-tubes, as well as most other brands of cryogenic vials. The reinforced seal, triple hinges and locking clasp keeps samples secure while the box is closed. Each box features and alpha-numeric grid on the exterior and interior, stacking rings and a matte surface suitable for printing and labeling.
Choose from one of the most recent versions:
Certificates of Analysis (COA)