Key Documents
Z688096
E-Vac polypropylene bottle
with quick release coupling lid, capacity 4,000 mL
About This Item
UNSPSC Code:
41121800
NACRES:
NB.22
material
polypropylene bottle
polypropylene cap
round bottle
joint
joint threaded
feature
with cap
packaging
pack of 1 ea
manufacturer/tradename
Cole Parmer 1305038
capacity
4,000 mL
fitting
GL 18 connector (2 ports & level sensor)
General description
The E-Vac bottles can be autoclaved for 20 minutes at 121°C to prevent laboratory contamination. Base units are illuminated to allow visual inspection of waste levels in the bottle. Dual hydrophobic filters prevent liquids from reaching the pump housing and contaminating the system.
E-Vac is available with a 4-L polypropylene bottle or a 3-L safety glass bottle. Polypropylene and glass bottles offer lids equipped with quick-release tubing connectors for safe and convenient tubing connections; and level detection sensor that automatically shuts the pump off when the bottle is full. Polypropylene bottle also available with lid fitted with standard barbed tube fittings.
Choose from one of the most recent versions:
Certificates of Analysis (COA)