Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z687510

3D Biotek 3D Insert PCL scaffold

for 100 mm dish

Synonym(s):

3D, 3D Cell Culture

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
NB.22

description

polycaprolactone (PCL)

sterility

sterile

manufacturer/tradename

3D Biotek PCL3030-01

H

~1.5 mm , scaffold

fiber diam.

~300 μm

size

~300 μm , spacing

color

clear

pore size

569 μm
65.6 % porosity (+/- 0.9)

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

3D Biotek offers cutting edge 3D cell culture products and technology at affordable prices, that are easy and convenient to use our 3D 3D Insert cell culture scaffolds are:

  • Terminally sterilized by gamma irradiation and ready for use
  • Compatible with most of your current 2D assays
  • Easy for imaging. Cell growth can be monitored by light microscope, no need to purchase any additional sophisticated equipment.

Legal Information

3D Insert is a trademark of 3D Biotek, LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Carlos E Caicedo-Carvajal et al.
Journal of tissue engineering, 2011, 362326-362326 (2011-11-11)
Isolation and amplification of primary lymphoma cells in vitro setting is technically and biologically challenging task. To optimize culture environment and mimic in vivo conditions, lymphoma cell lines were used as a test case and were grown in 3-dimension (3D)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service