Key Documents
Z683078
Plas-Labs automatic anaerobic chamber AC input 230 V
AC/DC input 230 V AC, UK plug
Synonym(s):
anaerobic chambers, humidity chambers, humidity control chambers, isolation chambers, nitrogen dry box
About This Item
UNSPSC Code:
41100000
description
includes glove box, shelving unit and support cart
manufacturer/tradename
Plas-Labs Simplicity-888
General description
The Plas-Labs automatic anaerobic chamber has a single station which is designed for automatic atmosphere control when working with oxygen sensitive materials, product containment, and general isolation control. This chamber features an internal PC with multifunctional operator interface.
  • Three drying train canisters with quick disconnects
  • Programmable catalyst heater unit
  • Bright fluorescent light system
  • White ambidextrous Hypalon® gloves
  • Two 1/4 in. NPT gas connections
  • Two ground key cock valves on main chamber
  • Electrical outlet strip; UL & CSA approved, hospital grade with four outlets
  • CE compliant
Other Notes
Price includes shipping
Legal Information
Hypalon is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)