Key Documents
Z667218
Greiner Easy-Load 1000 refill system
volume 100-1000 μL, blue, sterile
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
Pricing and availability is not currently available.
material
blue
polypropylene
sterility
sterile
packaging
pack of 2160 ea
manufacturer/tradename
Greiner 741045
availability
available only in EU
maximum volume
1 mL
volume
100-1000 μL
color
blue
General description
Ideally suited for Anthos, Brand, Costar, Eppendorf 4810, 4910, Eppendorf Reference, Finnpipette Digital, Gilson, Socorex Calibra single channel pipettors.
Legal Information
Easy-Load is a trademark of Cole-Parmer Instrument Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)