Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z666270

Greiner PS inoculating loop

size 10 μL, blue

Synonym(s):

inoculating needles, Greiner PS inoculating loop

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NB.22
Pricing and availability is not currently available.

material

polystyrene

sterility

sterile

packaging

pack of 60 ea

manufacturer/tradename

Greiner 731171

availability

available only in EU

size

10 μL

color

blue

Looking for similar products? Visit Product Comparison Guide

General description

Note: Disposable inoculating loop; hydrophobic

Legal Information


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service