Key Documents
Z659878
Bandelin SONOREX Ultrasonic bath accessories
tank lid for Z659843 and Z659053
About This Item
UNSPSC Code:
41103706
NACRES:
NB.74
Pricing and availability is not currently available.
material
stainless steel
manufacturer/tradename
Bandelin D 1028
availability
not available in USA and Canada
General description
When using chemically aggressive liquids, removing chemically aggressive or abrasive contamination or sonicating sample liquids, insert cups are used (indirect sonication). Because nothing may be placed on the bottom of the ultrasonic bath, use a suitable perforated lids/insert strips to hold the insert cups. Several insert cups can be sonicated with different liquids at the same time.
Features and Benefits
  • for holding insert cups
  • sound-absorbing
  • shorten the heating phase
Legal Information
SONOREX is a trademark of Bandelin Electronic GmbH & Co. KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)