Key Documents
Z659428
Bandelin SONOPULS ultrasonic homogenizer accessories
Indirect processing microtube holder EH 6
Synonym(s):
reaction cup holder
About This Item
UNSPSC Code:
41101520
Pricing and availability is not currently available.
packaging
pack of 1 ea
manufacturer/tradename
Bandelin 7503
availability
available only in EU
General description
For use in BB 6 (Z65,940-1). Up to 6 samples can be treated simultaneously. Pressure plate holds tubes in place. No floating of cups.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Mahmood Sinaei et al.
Ecotoxicology and environmental safety, 164, 675-680 (2018-09-01)
In this study, selected biomarkers-Phytochelatin (PC) and Metallothionein (MT)- were measured in the brown algae (i.e., Cystoseria indica) obtained from the Gulf of Oman. Chemical analyses were performed to measure the concentration of heavy metals (Cd, Cu, Zn, Pb, Hg