Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z659193

Bandelin SONOPULS ultrasonic homogenizer probes

TT 25 flat tip for use with HD 2200 homogenizer and SG 219G horn

Synonym(s):

flat probe sonicator, ultrasonic probe

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.74
Pricing and availability is not currently available.

material

titanium (Ti-6AI-4V alloy)

manufacturer/tradename

Bandelin

availability

available only in EU

diam.

25 mm

volume

30-1,000 mL , HD 2200

Looking for similar products? Visit Product Comparison Guide

General description

For use with HD 2200 homogenizer and SG 219G horn.
Transmit mechanical longitudinal waves into the sample. They are thermoresistant, can be treated in autoclaves and are resistant to corrosive media. Sample volume, diameter of the processing vessel and the required amplitude determine the selection of the unit and the type of probe. The higher the amplitude, the more intense the sonication.

Using the product image (from left to right):

MS 72 micro tip
MS 73 micro tip
KE 76 tapered tip
VS 70 T extended probe
VS 200 T extended probe
TT 13 flat tip
TT 19 flat tip
TT 25 flat tip

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service