Z657530
Eppendorf® IsoTherm-System®
IsoRack for 0.5mL Safe-Lock microcentrifuge tubes
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41100000
NACRES:
NB.22
feature
Contents: 4 racks plus 4 covers
manufacturer/tradename
Eppendorf® 3880 000.038
Looking for similar products? Visit Product Comparison Guide
General description
The IsoTherm System specializes in cooling samples effectively and constantly at -21 °C or 0 °C over many hours. It consists of the following components: the IsoRack, IsoPack and IsoSafe. It can be used as an enzyme bar to prevent enzymes from becoming inactive as well as for PCR sample preparation, sample transport and slow thawing of frozen samples.
- Transport and slow thawing of deep-frozen samples
- Holds 24 tubes (Safe-Lock 0.5 mL/1.5 mL/2.0 mL)
- IsoPack (cold pack), white for 0 °C and blue for -21 °C, cooled to refrigeration temperature in freezer compartment
- Isothermal times of < 6 h (0 °C, white) and < 3 h (-21 °C, blue)
- No more melting ice baths (danger of contamination)
- All system components can be used separately
- Racks stackable, autoclavable, markable, floatable and can be centrifuged in microtiter-plate rotor
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
IsoTherm-System is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service