Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z657514

Eppendorf® IsoTherm-System®

IsoRack + IsoPack 0°C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
NB.22

feature

Contents: 1 rack and 1x0.5mL cold pack

manufacturer/tradename

Eppendorf® 3880 000.160

Looking for similar products? Visit Product Comparison Guide

General description

The IsoTherm System specializes in cooling samples effectively and constantly at -21 °C or 0 °C over many hours. It consists of the following components: the IsoRack, IsoPack and IsoSafe. It can be used as an enzyme bar to prevent enzymes from becoming inactive as well as for PCR sample preparation, sample transport and slow thawing of frozen samples.
  • Transport and slow thawing of deep-frozen samples
  • Holds 24 tubes (Safe-Lock 0.5 mL/1.5 mL/2.0 mL)
  • IsoPack (cold pack), white for 0 °C and blue for -21 °C, cooled to refrigeration temperature in freezer compartment
  • Isothermal times of < 6 h (0 °C, white) and < 3 h (-21 °C, blue)
  • No more melting ice baths (danger of contamination)
  • All system components can be used separately
  • Racks stackable, autoclavable, markable, floatable and can be centrifuged in microtiter-plate rotor

Application

Eppendorf® IsoTherm-System® has been used to cool down samples during hydrolase assays.[1]

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
IsoTherm-System is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Industrial by-products from white-rot fungi production. Part I: Generation of enzyme preparations and chemical, protein biochemical and molecular biological characterization.
Schimpf U and Schulz R
Process. Biochem., 51, 2034-2046 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service