Key Documents
Z655368
Hanna combination pH electrodes - application specific
open junction, viscolene - micro electrode for 96 well plates
About This Item
UNSPSC Code:
41115603
description
single, Ag/AgCI
manufacturer/tradename
Hanna HI 1083B
pH measuring range
0-13
parameter
0-50 °C temp. range
0.1 bar max. pressure
General description
Glass body, Spherical tip shape (diam. 3 mm)
Help in selecting the correct electrode can be viewed at Electrode Selection Guide
Hints on Electrode Maintenance
Choose from one of the most recent versions:
Certificates of Analysis (COA)