Z654620
Sartorius eLINE® multichannel electronic pipettor
volume 10-300 μL, 8 channel, AC/DC input 110 V / 230 V AC, AC/DC input, universal plug set
Synonym(s):
Biohit pipettes, electronic pipettes, pipettor
About This Item
Recommended Products
feature
CE compliant
electronic
manufacturer/tradename
Sartorius 730361
volume
10-300 μL
General description
- Forward pipetting with optional cycle counting
- Reverse and manual pipetting modes
- Multi-dispensing; Diluting
- Automated Dispensing
- Mixing facility
The eLINE can store up to 6 user routines guaranteeing consistency of results irrelevant of the intricacy of the task undertaken. Tip ejection can be a chore at best and at worst a contributing factor to work-related upper limb disorders.
The eLINE negates this with its revolutionary electronic ejection system which discards the used tip at a touch of a button. Sartorius eLINE Electronic Pipettors should be charged using the original Sartorius eLINE Charging Stand. They also feature direct charging by using the Sartorius eLINE AC-adaptor.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)