Key Documents
Z654566
Sartorius eLINE® single channel electronic pipettor
volume 0.2-10 μL, AC/DC input 230 V AC, universal plug set
Synonym(s):
Biohit pipettes, electronic pipettes, pipettor
About This Item
UNSPSC Code:
41121500
feature
CE compliant
electronic
single-channel
packaging
pack of 1 ea
manufacturer/tradename
Sartorius 730021
volume
0.2-10 μL
battery power
rechargeable
General description
Sartorius eLINE electronic pipettors of the Biohitfamily offer the latest technology in liquid handling. The eLINE electronic pipettor has been designed to deliver the highest levels of analytical accuracy with minimal the eLINE pipettors together with the ejection - take the effort out of pipetting in every laboratory.
The eLINE modes including:
  • Forward Pipetting with Optional Cycle
  • Reverse Pipetting
  • Manual Pipetting
  • Post Delivery Mixing
  • Multi-Dispensing
  • Multi-Aspirating
  • Automated Multi-Dispensing
  • Sequential-Dispensing
Supplied with a universal adapter for Euro, UK and China plugs.
Legal Information
eLINE is a registered trademark of Sartorius Biohit Liquid Handling Oy
Choose from one of the most recent versions:
Certificates of Analysis (COA)