Key Documents
Z654493
Vilber Lourmat Electronic Ballast transilluminators
ETX-F20.M
About This Item
Pricing and availability is not currently available.
AC/DC input
110 / 230 V, 50-60 Hz
manufacturer/tradename
Vilber Lourmat 2111 2002 1
availability
available only in EU
table size
20 cm × 20 cm
General description
These 312 nm UV transilluminators are based on an electronic ballast developed by Vilber Lourmat. Available as 8 watt (ECX models) or 15 watt (ETX models). It′s 25,000 Hz frequency provides a supreme light stability and avoids the usual flickering effect. No cooling fan is necessary, as the electronic ballast produces no heat. All models have an adjustable safety screen and stainless steel frame.
  • Compact size
  • Reduced electrical consumption
Choose from one of the most recent versions:
Certificates of Analysis (COA)