Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z654043

Eppendorf® twin-tec PCR plates, 384-wells

colorless

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
NACRES:
NB.22
material:
colorless wells
conical bottom wells
polypropylene
size:
384 wells
sterility:
non-sterile
feature:
autoclavable
lid: no
skirt
Product Z654043 is not currently sold in your country. Contact Technical Service

material

colorless wells
conical bottom wells
polypropylene

sterility

non-sterile

feature

autoclavable
lid: no
skirt

packaging

pack of 25 ea

manufacturer/tradename

Eppendorf® 0030 128.508

availability

available only in EU

technique(s)

PCR: suitable

size

384 wells

well maximum volume

40 μm

well volume

250 μL

working volume

250 μL

color

colorless

Looking for similar products? Visit Product Comparison Guide

General description

Eppendorf twin-tec PCR plates give you the advantages that come from combining two materials: the wells are manufactured from virgin polypropylene, the most successful material used for PCR tubes; polycarbonate, an especially rigid material, forms the plate deck and the skirt. This one-piece design is achieved by an innovative injection moulding process, twin-shot technology.
  • Minimal bending of skirts
  • Semi-skirted for labeling and bar-coding
  • Suitable for quantitative real-time PCR
  • Fits most Thermal Cyclers
  • 250 μL max. well volume (when used with cap strips)

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service