Key Documents
Z653551
Pyrex® glass test tubes without rim, medium wall
O.D. × L 24 mm × 150 mm, wall thickness 1.2 mm
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Pricing and availability is not currently available.
material
borosilicate glass
clear tube
packaging
pack of 100
manufacturer/tradename
Bibby Pyrex 1622/20M
O.D. × L
24 mm × 150 mm
wall thickness
1.2 mm
General description
  • Rimless for greater convenience in plugging and rack storage
  • Ideal for culture work
  • Fully autoclavable
  • High resistance to attack by water
  • Reduces leaching of contaminants which can cause pH changes
  • Manufactured from heat resistant Pyrex borosilicate glass allowing use at high temperatures
  • Conforms to ISO 4142
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)