Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z651494

Eppendorf® twin-tec PCR plates 96, semi-skirted

red

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
NACRES:
NB.22
Pricing and availability is not currently available.

material

polycarbonate
red wells

description

Colorless wells

feature

autoclavable
skirt (Semi skirted)

packaging

pack of 1 ea

manufacturer/tradename

Eppendorf® 0030 128.613

availability

available only in EU

technique(s)

PCR: suitable

size

96 well

well maximum volume

250 μm

color

red

Looking for similar products? Visit Product Comparison Guide

General description

Eppendorf twin-tec PCR plates give you the advantages that come from combining two materials: the wells are manufactured from virgin polypropylene, the most successful material used for PCR tubes; polycarbonate, an especially rigid material, forms the plate deck and the skirt. This one-piece design is achieved by an innovative injection moulding process, twin-shot technology.
  • Minimal bending of skirts
  • Semi-skirted for labeling and bar-coding
  • Suitable for quantitative real-time PCR
  • Fits most Thermal Cyclers
  • 250 μL max. well volume (when used with cap strips)

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service