Z651451
Eppendorf® twin-tec PCR plates 96, semi-skirted
yellow
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106300
NACRES:
NB.22
material
colorless wells
polycarbonate
description
Colorless wells
feature
autoclavable
skirt (Semi skirted)
packaging
pack of 25 ea
manufacturer/tradename
Eppendorf® 0030 128.583
availability
available only in EU
technique(s)
PCR: suitable
size
96 well
working volume
250 μL
color
yellow
Looking for similar products? Visit Product Comparison Guide
General description
Eppendorf twin-tec PCR plates give you the advantages that come from combining two materials: the wells are manufactured from virgin polypropylene, the most successful material used for PCR tubes; polycarbonate, an especially rigid material, forms the plate deck and the skirt. This one-piece design is achieved by an innovative injection moulding process, twin-shot technology.
- Minimal bending of skirts
- Semi-skirted for labeling and bar-coding
- Suitable for quantitative real-time PCR
- Fits most Thermal Cyclers
- 250 μL max. well volume (when used with cap strips)
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service