Key Documents
Z649872
Sigma-Aldrich
Additional or replacement plates for TV100 user-cast gels
Plain glass plate 100 × 100 mm, bonded 1.0 mm spacers
About This Item
UNSPSC Code:
12352200
NACRES:
NC.01
Pricing and availability is not currently available.
availability
available only in EU
General description
Note: TV100 is a dual unit and typically cast with 2 gels on module. Each user-cast gel requires 1 notched plate and one plain plate. Pack = 2 plates.
Choose from one of the most recent versions:
Certificates of Analysis (COA)