Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z649767

Sigma-Aldrich

TV100Y mini vertical electrophoresis unit for pre-cast and user-cast gels

Tank and gel running module with casting accessories

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
NC.01
Pricing and availability is not currently available.

grade

electrophoresis grade

feature

Buffer volume: 400 mL min - 1200 mL max

availability

available only in EU

outside size

20 cm × 15.5 cm × 14.5 cm

plate size

10 cm × 10 cm

Looking for similar products? Visit Product Comparison Guide

General description

  • Improved design for pre-cast gels
  • Improved parallel clamping system prevents lane distortion
  • Compatible pre-cast gels 10 × 10 cm, most pre-cast gels 8 × 10 cm
  • Large buffer volume for complete immersion of gels
  • Casting base and accessories for user-cast gels
  • Improved seals when casting

Suitable for Protein and Nucleic Acid separations, Mini SDS-PAGE, Preparative, Peptide and Gradient Pore separations.
Dual gel unit, designed to meet the growing demand for running pre-cast polyacrylamide gels and including all the accessories needed for user to cast own gels. Easy use, wide-jaw, quick-release parallel clamping system that accepts most gels and applies even pressure onto cassettes or plates, preventing lane distortion from physical strain. Gels are fully submerged in running buffer, evenly distributing heat to reduce band distortion. Safety lid and asymmetric offset electrodes prevent accidental polarity reversal.
Casting accessories included: 2 × 2 mm notched glass plates, 2 × 2 mm plain glass plates bonded with 1 mm spacers, 1 × dummy plate, 2 × 1 mm 12 sample well combs and casting base with silicon seals.
Plain glass plates have bonded spacers to facilitate assembly and casting of gels, bonded spacers correctly positioned for simple, easy casting
Wide range of combs listed with Mini Vertical Electrophoresis unit may be used with the casting accessories in this set.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service