Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z648043

Millipore

Amicon Ultra centrifugal filter units

Ultra-4, MWCO 100 kDa

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103900
NACRES:
NB.24

material

polyethylene cap
polypropylene tube
regenerated cellulose membrane
styrene-butadiene housing

sterility

non-sterile

manufacturer/tradename

Millipore UFC810024

concentrate volume

50 μL

membrane area × diam. × L

3 cm2 × 22.7 mm × 124 mm

tube size

22.7 x 124 mm

working volume

~4 mL

pore size

100 kDa MWCO

Looking for similar products? Visit Product Comparison Guide

General description

High recovery centrifugal filters for concentrating samples. Can be used to concentrate biological samples containing antigens, antibodies, enzymes, nucleic acids or micoorganisms. Excellen for use to purify macromolecular components from cell lysates. Ideal for desalting and buffer exchanges.
Swing bucket rotorL 4,000 x g; Fixed angle rotor (35°): 5,000 x g.
  • High recovery Ultracel regenerated cellulose membrane in a range of molecular weight cut-offs.
  • High retentate recovery of >90%
  • Vertical membrane reduces concentration polarization for ultra-fast spin times (as fast as 10-15 mins)
  • Heat-sealed membrane minimizes downstream extractables
  • 100% integrity tested for reliable performance
  • Convenient sample monitoring with translucent housing and volume graduations
  • Direct pipettor sample access eliminates processing step to recover concentrate
  • High concentration factors of 80 - 100X

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laura A Powell et al.
Cell host & microbe, 28(5), 699-711 (2020-08-14)
Mosquito inoculation of humans with arthritogenic alphaviruses results in a febrile syndrome characterized by debilitating musculoskeletal pain and arthritis. Despite an expanding global disease burden, no approved therapies or licensed vaccines exist. Here, we describe human monoclonal antibodies (mAbs) that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service