Key Documents
Z646687
BRAND® Transferpette® S starter kits
Micro
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
manufacturer/tradename
BRAND 704790
General description
Contains a set of 3 Transferpette S, Digital adjustable volume, 3 filled Tip-Box N and 3 wall/rack mounts.
Includes Transferpette S models D-1, D-10 & D-100
Features and Benefits
  • Efficient: one-hand operation
  • Flexible: ergonomic design for all hand sizes
  • Reliable: volume-change protection prevents accidental adjustments
Other Notes
BR704790 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Transferpette is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)