Key Documents
Z645567
Nichiryo pipette tips
volume range 0.1-10 μL, elongated, non-sterile
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
volume:
0.1—10 μL
sterility:
non-sterile
feature:
barrier: no
packaging:
pack of 960 ea (10 racks of 96 tips)
manufacturer/tradename:
Nichiryo BMT-SSR
material
colorless
sterility
non-sterile
feature
barrier: no
packaging
pack of 960 ea (10 racks of 96 tips)
manufacturer/tradename
Nichiryo BMT-SSR
availability
not available in (France)
maximum volume
10 μL
volume range
0.1-10 μL
volume
0.1—10 μL
Features and Benefits
  • RNase and DNase free
  • Fits most pipettes
  • Premium packaging featuring stand-up and puncture-resistant bags
  • Racked tips are offered in both hinged and lift-off tops
Other Notes
Browse all pipette tips
Choose from one of the most recent versions:
Certificates of Analysis (COA)