Key Documents
Z640859
Rocking table Kick II
platform
About This Item
UNSPSC Code:
41100000
NACRES:
NC.01
Pricing and availability is not currently available.
feature
CE compliant
availability
available only in EU
General description
Rocking table KICK II, thermostattable large size incubation platform suitable for various incubation and preparation techniques (e. g. ELISAs, immunodiffusion, immunoblotting, histochemistry, in-situ-hybridization), cell- and microbiological culture experiments and for the accelerated processing of staining/destaining procedures with electrophoretic gels and blotting membranes.
A rugged and robust heavy-duty lab instrument with loading capacity up to 5 kg and an effective area of 45 × 30 cm covered with an anti-slip support. Constructed for continuous operation. Electronic regulation of lifting frequency (10 - 100/min) with newly developed lifting technique (lifting angle 5°, lifting height 3 cm) performing a continuous light "kick" agitation to prevent gels and membranes from sticking to their containment support!
Choose from one of the most recent versions:
Certificates of Analysis (COA)