Z640700
Uniphor electrophoresis units
S-80/120, universal horizontal unit
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
NACRES:
NC.01
Recommended Products
grade
electrophoresis grade
manufacturer/tradename
S-80/120
electrode distance
≥4 mm × 100 mm
gel size
≤80 mm × 120 mm
weight
1.6 kg
output
~3000 V, ≤25 mA
Looking for similar products? Visit Product Comparison Guide
General description
UniPhor units can be used for:
Analytical Protein and Peptide Characterization: Denaturing and non denaturing SDS-PAGE gels IEF in ultrathin PAGE gels (Z640727, Z640719 & Z640751)
Preparative Protein and Peptide Separation: Non-denaturating PAGE gels and Agarose gels IEF in Agarose Ampholyte Gels (Z640719 & Z640751)
Molecular Biology: DNA/RNA fragment analysis, SSCP-Electrophoresis, Long Distance-TGGE (needs external cryo-thermostate with temperature ramping functionality), Flat-Bed Sequencing (Z640743 & Z640751).
All units are equipped with SCE (Solid Carbon Electrodes) electrodes, which help avoid any of the known problems of Platinum wire electrodes due to their higher conductivity and larger contact area with the gel.
Analytical Protein and Peptide Characterization: Denaturing and non denaturing SDS-PAGE gels IEF in ultrathin PAGE gels (Z640727, Z640719 & Z640751)
Preparative Protein and Peptide Separation: Non-denaturating PAGE gels and Agarose gels IEF in Agarose Ampholyte Gels (Z640719 & Z640751)
Molecular Biology: DNA/RNA fragment analysis, SSCP-Electrophoresis, Long Distance-TGGE (needs external cryo-thermostate with temperature ramping functionality), Flat-Bed Sequencing (Z640743 & Z640751).
All units are equipped with SCE (Solid Carbon Electrodes) electrodes, which help avoid any of the known problems of Platinum wire electrodes due to their higher conductivity and larger contact area with the gel.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service