Z640379
Eppendorf® epT.I.P.S. set
volume range 500-2500 μL
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
volume:
500—2500 μL
sterility:
non-sterile
feature:
barrier: no
packaging:
pack of 240 (5 trays of 48 tips, includes 1 reusable box)
manufacturer/tradename:
Eppendorf® 0030 073.347
Eppendorf® 022491474
Eppendorf® 022491474
Recommended Products
material
red tip
sterility
non-sterile
feature
barrier: no
packaging
pack of 240 (5 trays of 48 tips, includes 1 reusable box)
manufacturer/tradename
Eppendorf® 0030 073.347
Eppendorf® 022491474
L
115 mm
maximum volume
2.5 mL
volume range
500-2500 μL
volume
500—2500 μL
Looking for similar products? Visit Product Comparison Guide
General description
Part of the ep T.I.P.S. box and reload system for contamination-free handling of refill trays, with plastic lids protecting the trays from contact during transfer to the working box.
Autoclavable working box, optimized for multichannel pipettes, with removable lid and larger footprint to avoid tipping, supplied with 5 trays of tips.
Autoclavable working box, optimized for multichannel pipettes, with removable lid and larger footprint to avoid tipping, supplied with 5 trays of tips.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service