Key Documents
Z640344
Eppendorf® epT.I.P.S. set
volume range 20-300 μL
Synonym(s):
pipetting tips
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
volume:
20—300 μL
sterility:
non-sterile
feature:
barrier: no
packaging:
pack of 480 (5 trays of 96 tips, includes 1 reusable box)
manufacturer/tradename:
Eppendorf® 0030 073.282
Eppendorf® 022491440
material
orange tip
sterility
non-sterile
feature
barrier: no
packaging
pack of 480 (5 trays of 96 tips, includes 1 reusable box)
manufacturer/tradename
Eppendorf® 0030 073.282
Eppendorf® 022491440
L
55 mm
maximum volume
300 μL
volume range
20-300 μL
volume
20—300 μL
General description
Part of the ep T.I.P.S. box and reload system for contamination-free handling of refill trays, with plastic lids protecting the trays from contact during transfer to the working box.
Autoclavable working box, optimized for multichannel pipettes, with removable lid and larger footprint to avoid tipping, supplied with 5 trays of tips.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)