Key Documents
Z640212
Eppendorf® epT.I.P.S. box
volume range 0.5-20 μL
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
volume:
0.5—20 μL
sterility:
non-sterile
feature:
barrier: no
packaging:
pack of 96 (includes 1 reusable box)
manufacturer/tradename:
Eppendorf® 0030 073.045
Eppendorf® 022491326
material
light gray tip
sterility
non-sterile
feature
barrier: no
packaging
pack of 96 (includes 1 reusable box)
manufacturer/tradename
Eppendorf® 0030 073.045
Eppendorf® 022491326
L
46 mm
maximum volume
20 μL
volume range
0.5-20 μL
volume
0.5—20 μL
General description
Part of the ep T.I.P.S. box and reload system for contamination-free handling of refill trays, with plastic lids protecting the trays from contact during transfer to the working box.
Autoclavable working box, optimized for multichannel pipettes, with removable lid and larger footprint to avoid tipping. Stackable. Supplied full of tips.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)