Key Documents
Z640093
Eppendorf® epT.I.P.S. standard
volume range 2-200 μL
Synonym(s):
pipetting tips
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
volume:
2—200 μL
sterility:
non-sterile
feature:
barrier: no
packaging:
pack of 1000 ea (2 bags of 500 tips)
manufacturer/tradename:
Eppendorf® 0030 000.870
Eppendorf® 022492039
material
yellow tip
sterility
non-sterile
feature
barrier: no
packaging
pack of 1000 ea (2 bags of 500 tips)
manufacturer/tradename
Eppendorf® 0030 000.870
Eppendorf® 022492039
L
53 mm
maximum volume
200 μL
volume range
2-200 μL
volume
2—200 μL
General description
Bulk packed in re-sealable bags.
200 μL, 300 μL and 1000 μL tips are color coded, yellow, orange and blue respectively.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)