Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z637335

Pursept®-A disinfectant

5 L canisters

Synonym(s):

Antimicrobial Solution, Surface Disinfectant

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47131800
NACRES:
NB.35
Pricing and availability is not currently available.

manufacturer/tradename

BRAND 44835

application(s)

detection

Looking for similar products? Visit Product Comparison Guide

General description

Ingredients: 100 g Pursept® AF contain: 12.5 g didecyl-dimethyl-ammoniumchloride, 10.4 g alkylpropylenediaminebis-guanidiniumdiacetate, 1.5 g bis(aminopropyl)-laurylamine, 5 - 15% NTA, < 5% non-ionic surfactants, auxiliary agent.

Features and Benefits

  • Suitable for disinfecting sensitive communication equipment
  • Dermatologically tested
  • Ready for use

Other Notes

BR44835 (India)

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG
Pursept is a registered trademark of Merz GmbH & Co. KG

pictograms

FlameExclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

73.4 °F

flash_point_c

23 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service