Z629391
Vivaspin 500 centrifugal concentrators
pore size 0.2 μm
About This Item
Recommended Products
material
polycarbonate housing
polyethersulfone membrane
sterility
non-sterile
feature
holdup volume 5 μL
packaging
package of 100 ea
manufacturer/tradename
Sartorius VS0172
availability
not available in ROW
parameter
15000 × g max. centrifugal force
dead stop volume
5 μL
membrane area
0.5 cm2
tube size
~2.2 mL
working volume
500 μL
pore size
0.2 μm
Looking for similar products? Visit Product Comparison Guide
General description
- Vivaspin centrifugal concentrators offer the optimal solution to any concentration or buffer exchange application with their broad range of volume capacities and MWCOs.
- Highest flow rates and recoveries are achieved due to their particular vertical membrane design.
- Incomparably safe sample handling and convenience is assured by the patented dead-stop technology allowing direct recovery of the concentrate without further re-spin.
Note: Vivaspin 500 μL centrifugal filter units offer a simple one step procedure for sample preparation. They can effectively be used in either swing out or fixed angle rotors.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service