Z629294
Vivacell 250 ultrafiltration concentrator
Complete with pressure head, pressure indicator/over pressure
All Photos(1)
About This Item
UNSPSC Code:
41104916
NACRES:
NB.24
Recommended Products
material
polycarbonate housing
polyethersulfone membrane
sterility
non-sterile
packaging
package of 1 ea
manufacturer/tradename
Sartorius VCA250
availability
not available in ROW, USA and Canada
volume measuring range
50-250 mL
working volume
250 mL
General description
The Vivacell 250 is a totally new concept for the concentration of larger biological samples. This product offers numerous advantages when compared to stirred cells.
- One size handles a volume range from under 50 mL to 250 mL.
- The easy-to-fit membrane insert enables rapid change of membrane.
- Use free standing on a bench top or in a refrigerator for maximum simplicity, or use on laboratory shaker for fastest concentrations.
- The unique conical dead stop built into the bottom of the membrane insert allows concentrations to under 1 mL.
- The gentle vortex action controls membrane polarisation whilst greatly reducing the shear effects typical of stirring mechanisms.
Features and Benefits
Includes pressure head, pressure indicator/over pressure release valve, 2 sample reservoirs and filtrate vessels, quick release connections to APC.
Choose from one of the most recent versions:
Certificates of Analysis (COA)