Key Documents
Z627925
Sigma-Aldrich
BRAND® EASYCAL 4.0 calibration software
Basic version
Select a Size
1 EA
$412.00
$412.00
Check Cart for Availability
Select a Size
Change View
1 EA
$412.00
About This Item
UNSPSC Code:
41100000
NACRES:
NA.22
$412.00
Check Cart for Availability
General description
EasyCal 4.0 calibration software performs all calculations automatically, matches them to standards and generates a report to document the calibration. All results are stores in a database for long term analysis. Individual testing intervals can be defined.
Available as basic version (manual entry of measured values) or as Professional version with electronic transfer of gravimetric data from balance via RS interface.
Features and Benefits
  • Simple: can be used with pipettes and instruments of all manufacturers.
  • Easy: Automatic calculation using the up-to-date standards.
  • Efficient: Professional and transparent recording of all parameters.
Other Notes
BR708445 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
EASYCAL is a trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Protocols
This article provides testing instructions for BRAND Dispensette® S bottle-top liquid dispenser according to the standard ISO DIS 8655.