Key Documents
Z627585
BRAND® QuikSip BT-Aspirator
complete kit
About This Item
UNSPSC Code:
41121800
NACRES:
NB.22
grade
CE Compliant
feature
Items supplied: 1 QuikSip
BT-Aspirator, 1 cell culture unit, 1 spare dispensing cartridge, 2 PP adapters (GL 45/32 and GL 45/S40)
manufacturer/tradename
BRAND
General description
Hand-held aspirator is designed for the safe removal of supernatants, such as culture media from dishes, flasks, culture bottles and multiwell plates or from supernatants after precipitation and centrifugation of proteins and nucleic acids.
No external vacuum source is required. For fingertip vacuum control, the cell culture unit is connected to the QuikSip BT-Aspirator.
A single channel device for removal of liquids out of reaction vessels, tubes etc is supplied as standard. Suitable for use with disposable pipette tips, micropettes and glass Pasteur pipettes.
Other Notes
BR4723150 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
QuikSip is a trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)