Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z617709

Greiner Sapphire PCR tube caps

0.2 mL, thin wall, PCR strip caps only

Synonym(s):

microcentrifuge tubes, microfuge tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.11

material

domed cap

feature

RNase and DNase free

manufacturer/tradename

Greiner 373270

packaging

case of 8 caps/strip (10x125 strips)

technique(s)

PCR: suitable

General description

Caps only. Tubes are not included.
Greiner PCR tubes′ thin wall construction promotes optimal heat transfer between the thermocycler block and the reaction mixture. These PCR tubes are certified RNase/DNase free and are non-pyrogenic as well.

Legal Information


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service