Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z617369

Greiner pipette tips

maximum volume 100 μL, non-sterile, pack of 500 ea

Synonym(s):

Greiner pipette tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
NACRES:
NB.01

description

for Eppendorf Reference & Research pipettes

sterility

non-sterile

feature

graduated: no

packaging

pack of 500 ea

manufacturer/tradename

Greiner 685290

maximum volume

100 μL

color

yellow

Looking for similar products? Visit Product Comparison Guide

Other Notes

Browse all pipette tips

Legal Information


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service