Z614718
Vivaspin 20 centrifugal concentrator
pore size 0.2 μm
About This Item
material
polycarbonate housing
polyethersulfone membrane
description
PES membrane
sterility
non-sterile
feature
holdup volume 20 μL
packaging
package of 48 ea
manufacturer/tradename
Sartorius VS2072
parameter
5 bar max. pressure
8000 × g max. centrifugal force
dead stop volume
50 μL
membrane area
6 cm2
tube size
~50 mL
working volume
20 mL
pore size
0.2 μm
Looking for similar products? Visit Product Comparison Guide
General description
Remaining volume is easy to read off the printed scales on the side of the concentrator and the modified dead stop pocket further simplifies direct pipette recover of the final concentrate.
- Vivaspin centrifugal concentrators offer the optimal solution to any concentration or buffer exchange application with their broad range of volume capacities and MWCOs
- Highest flow rates and recoveries are achieved due to their particular vertical membrane design
- Incomparably safe sample handling and convenience is assured by the patented dead-stop technology allowing direct recovery of the concentrate without further re-spin
Note: One step dealing of concentrated sample using diafiltration cup. Gas pressure filtration; for faster processing gas pressure can be combined with centrifugal force.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service