Key Documents
Z613789
Whatman® Nytran SuPerCharge (SPC) nylon blotting membranes
0.45 μm nylon blotting membranes 82 mm circles 50/pk
Synonym(s):
blotting membranes, nylon blotting membranes
About This Item
UNSPSC Code:
41100000
material
Nylon membrane (Nytran SuPerCharge (SPC))
packaging
pack of 50
manufacturer/tradename
Whatman 10416216
Whatman Article No. 28415365 (US reference)
diam.
82 mm
pore size
0.45 μm
suitability
suitable for (Southern and Northern blotting; colony and plaque lifts and Dot-/Slot-Blots)
General description
Nylon membrane providing consistently high signal with extremely low background. Nytran SuPerCharge (SPC) Blotting Membranes provide greater reproducibility of results across the membrane and from blot to blot.
Nytran SPC membrane is cast uniformly on both sides of a support matrix, demonstrating excellent symmetry. This gives the membrane the ability to lie flat without curling. With typical manufacturing techniques, increasing positive charge tends to increase background. Nytran SPC membranes are manufactured using a process that allows the combination of high positive charge with low background. Whether using radioactive or nonradioactive detection techniques, Nytran SPC consistently gives high signal with extremely low background. Whatman Nytran nylon membranes are available in two formats: Nytran N is moderately charged and Nytran SuPerCharge (SPC) has a very high positive charge.
Other Notes
Global SIAL customers (excluding Japan) can now order any product from the Cytiva Life Sciences consumables portfolio (prepack only) using GE′s product numbering system.
The legacy GE Whatman products that use the SIAL "Z" product numbering system will be discontinued and directly replaced with a product number using a "WHA" prefix plus the GE product code. For reference, the legacy "Z" product number is identified as a synonym on the "WHA" product number listing.
Legal Information
Nytran is a trademark of Cytiva
Whatman is a registered trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)